Learn More
Electron Microscopy Sciences Sodium Cacodylate Buffer, 0.4M, 500 mL
SDP
Catalog No. 50366184
Encompass_Preferred
Prepared from Sodium Cacodylate Trihydrate (F.W. 214.02), pH 7.2.
Catalog No. 50-366-184 Supplier Electron Microscopy Sciences Supplier No. 11654
Prepared from Sodium Cacodylate Trihydrate (F.W. 214.02), pH 7.2.