Learn More
Electron Microscopy Sciences Sodium Cacodylate Buffer, 0.4M, pH 7.2 1 LT
SDP
Catalog No. 50366664
Encompass_Preferred
Prepared from Sodium Cacodylate Trihydrate (F.W. 214.02). Our buffers are ideal for maintaining a constant pH during fixation and have an ionic constitution to prevent the extraction of cellular components or the deposition of precipitates of fixative chemicals. The buffers are prepared fresh for each order and conveniently packaged.
Standard pH: 7.2
Catalog No. 50-366-664 Supplier Electron Microscopy Sciences Supplier No. 11655
Prepared from Sodium Cacodylate Trihydrate (F.W. 214.02). Our buffers are ideal for maintaining a constant pH during fixation and have an ionic constitution to prevent the extraction of cellular components or the deposition of precipitates of fixative chemicals. The buffers are prepared fresh for each order and conveniently packaged.
Standard pH: 7.2