Learn More
Strem, An Ascensus Company CAS# 6131-99-3. 25g. Sodium cacodylate trihydrate, 98%. MFCD00149079
SDP
Supplier:  Strem, An Ascensus Company 11100025G
Encompass
CAS# 6131-99-3. 25g. Sodium cacodylate trihydrate, 98%. MFCD00149079. Molecular Weight: 159.91. Molecular Formula: (CH3)2As(O)ONa·3H2O. Color/form: white pwdr. Strem# 11-1000. http://www.strem.com/catalog/v/11-1000/
Catalog No. 50-901-09462