Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium Cacodylate Trihydrate

Catalog No. 20840500GF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-840-500GF 500 g
20-840-100GF 100 g
20-840-25GF 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 20-840-500GF Supplier Sigma Aldrich Supplier No. 20840500GF
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium cacodylate trihydrate is a biological buffer. Its crystals have been analyzed by single-crystal X-ray diffraction studies. Studies suggested its crystals exhibited two phases, viz. penta-++-aqua-disodium(I) bis(dimethylarsenate), {[Na2(H2O)5](C2H6AsO2)2}n and di-++-aqua-bis[triaquasodium(I)] bis(dimethylarsenate), [Na2(H2O)8](C2H6AsO2)2.Sodium cacodylate trihydrate may be employed as buffer to maintain pH 7.4 in electron microscopic studies. It may be used to compose cacodylate buffer for use in electron microscopic studies.

Specifications

CAS 6131-99-3
Molecular Formula C2H6AsNaO2 -+ 3H2O
Linear Formula (CH3)2AsO2Na -+ 3H2O
MDL Number MFCD00149079
Quantity 500 g
Synonym Cacodylic acid sodium salt trihydrate; Dimethylarsinic acid sodium salt; Dimethylarsonic acid sodium salt
Molecular Weight (g/mol) 214.03 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.