Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Caprate, MP Biomedicals

Catalog No. ICN221694 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN221694 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN221694 Supplier MP Biomedicals, Inc Supplier No. 0522169405
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1002-62-6
Molecular Formula C10H19NaO2
Molecular Weight (g/mol) 194.25
MDL Number MFCD00066453
InChI Key FIWQZURFGYXCEO-UHFFFAOYSA-M
Synonym sodium decanoate, sodium caprate, decanoic acid, sodium salt, decanoic acid sodium salt, sodium caprinate, capric acid sodium salt, sodium-n-decanoate, sodium decanoic acid, capric acid, sodium salt, caprinic acid sodium salt
PubChem CID 4457968
IUPAC Name sodium decanoate
SMILES [Na+].CCCCCCCCCC([O-])=O

Specifications

Quantity 5 g
Chemical Name or Material Sodium Caprate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.