Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Carbonate, 0.05 Normal Standardized,, Reagents

Catalog No. 01333774
Change view
Click to view available options
Quantity: 500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-333-774 Quantity: 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-333-774 Supplier Reagents Holdings Llc Supplier No. CS122500500A
 Restricted Item
Add to Cart
Add to Cart

Sodium Carbonate, 0.05 Normal (1ml = 2.5mg CaCO3) Standardized, NIST Traceable, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Sodium Carbonate
Boiling Point 100°C
Color Colorless
CAS 7732-18-5, 497-19-8
pH 10 to 11.5
Physical Form Liquid
Quantity 500 mL
Molecular Formula Na2CO3/H2O
Synonym Soda ash, Disodium carbonate
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.0016
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.