Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Carbonate, 0.0500 N (N/20), 1 mL = 2.5 mg CaCO3, Ricca Chemical
SDP

Catalog No. 718516 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle

Catalog No. 7185-16

Supplier: Ricca Chemical Company 718516

Only null left
Add to Cart
Add to Cart

APHA/EPA for Alkalinity Testing

Specifications

CAS 7732-18-5
Grade Laboratory
Packaging Natural Poly Bottle
Methods APHA/EPA for Alkalinity
pH 11 to 11.5
Quantity 500 mL
Chemical Name or Material Sodium Carbonate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.