Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Carbonate, 10% w/v, Reagents

Catalog No. 01333867
Change view
Click to view available options
Quantity:
500 mL
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-333-867 4 L
01-333-866 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-333-867 Supplier Reagents Holdings Llc Supplier No. CS1028004A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium Carbonate, 10% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 497-19-8
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Soda ash, Disodium carbonate
IUPAC Name water
SMILES O

Specifications

Boiling Point 100°C
Color Colorless
pH >12
Physical Form Liquid
Quantity 4 L
Percent Purity 10% w/v
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.0957
Chemical Name or Material Sodium Carbonate, 10% w/v
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.