Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Carbonate, 2.00 N, Ricca Chemical
SDP

Catalog No. 719116 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
719116 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 719116 Supplier Ricca Chemical Company Supplier No. 719116
Add to Cart
Add to Cart

Specifications

Color Colorless
CAS 497-19-8, 7732-18-5
CAS Min % 89.0, 9.4
CAS Max % 92.0, 9.78
Molecular Formula CNa2O3
MDL Number MFCD00003494
Synonym sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
SMILES [Na+].[Na+].[O-]C([O-])=O
IUPAC Name disodium carbonate
Molecular Weight (g/mol) 105.99
PubChem CID 10340
ChEBI CHEBI:29377
Grade Laboratory
Packaging Natural Poly Bottle
pH >12
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 2.00Normal
Chemical Name or Material Sodium Carbonate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.