Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Carbonate, 5% (w/v), Ricca Chemical
SDP

Catalog No. 715532 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
7155-32 1 L Natural Poly Bottle
7155-16 500 mL Natural Poly Bottle
7155-1 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 7155-32 Supplier Ricca Chemical Company Supplier No. 715532
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 497-19-8 , 7732-18-5
Molecular Formula CNa2O3
Molecular Weight (g/mol) 105.99
MDL Number MFCD00003494
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
Synonym sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
PubChem CID 10340
ChEBI CHEBI:29377
IUPAC Name disodium carbonate
SMILES [Na+].[Na+].[O-]C([O-])=O

Specifications

Color Colorless
CAS Min % 93.0, 4.67
CAS Max % 97.0, 4.86
Percent Purity 5%
Grade Laboratory
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
pH 11.5 to 12
Quantity 1 L
Physical Form Liquid
Concentration or Composition Notes 5% (w/v)
Chemical Name or Material Sodium Carbonate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.