Learn More
Aqua Solutions Sodium Carbonate 900 mM IC Concentrate, NIST Traceable (1L)
SDP
Catalog No. NC9887059
Encompass
Sodium Carbonate 900 mM IC Concentrate, NIST Traceable (1L)
Catalog No. NC9887059 Supplier Aqua Solutions Supplier No. VUL308
Sodium Carbonate 900 mM IC Concentrate, NIST Traceable (1L)