Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium carbonate, 99.5%, extra pure, anhydrous

Catalog No. p-3737352
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
Plastic Bottle
Plastic Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC123670010 1 kg Plastic Bottle
AC123670025 2.5 kg Plastic Bottle
AC123670100 10 kg Plastic Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC123670010 Supplier Thermo Scientific Chemicals Supplier No. 123670010
Only null left
Add to Cart

CAS: 497-19-8 | CNa2O3 | 105.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Adsorbent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 497-19-8
Molecular Formula CNa2O3
Molecular Weight (g/mol) 105.99
MDL Number MFCD00003494
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
Synonym sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
PubChem CID 10340
ChEBI CHEBI:29377
SMILES [Na+].[Na+].[O-]C([O-])=O

Specifications

Boiling Point 1600.0°C
Melting Point 851.0°C
Color White
Physical Form Fine Granular Powder
Quantity 1 kg
Assay Percent Range 99.5%
Linear Formula Na2CO3
Merck Index 15, 8731
Solubility Information Solubility in water: 22 g/100mL (20°C). Other solubilities: soluble in glycerol, insoluble in alcohol
IUPAC Name disodium;carbonate
Formula Weight 105.99
Percent Purity 99.5%
Grade Extra Pure
Loss on Drying 1% max. (300°C, 2 hrs)
Packaging Plastic Bottle
Chemical Name or Material Sodium carbonate, Anhydrous
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF exposed or concerned: Get med
EINECSNumber 207-838-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.