Learn More
Research Products International Corp Sodium Carbonate Anhydrous [Soda Ash], ACS Grade, 1 Kilogram

Supplier: Research Products International Corp S250251000.0
Substance Name: Soda Ash
CAS Number: 497-19-8
Molecular Formula: Na2CO3
Molecular Weight: 105.99
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.