Learn More
Specifications
Specifications
| Boiling Point | 1600°C |
| Melting Point | 854°C |
| Color | White |
| pH | 11.3 |
| Physical Form | Solid |
| Quantity | 10 kg |
| Formula Weight | 105.99 |
| Grade | Laboratory |
| Identification | Pass Test |
| Packaging | Poly Pail |
| Show More |
Safety and Handling
| Hazard Statement | WARNING! |
| Precautionary Statement | Emergency Overview Irritating to eyes and skin. May cause central nervous system effects. Hygroscopic. Wear personal protective equipment. Ensure adequate ventilation. Do not get in eyes, on skin, or on clothing. Use personal protective equipment. Do not breathe dust. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air. NFPA Health:2 Flammability:0 Instability:1 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.