Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium carbonate decahydrate

Catalog No. 577782250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-778-2250G 250 g
57-778-21KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 57-778-2250G Supplier Sigma Aldrich Supplier No. 577782250G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium carbonate decahydrate (Na2CO3.10H2O) can be used in the preparation of eutectic hydrate salts, which can be used to remove phase change materials for thermal energy storage. It can also be dehydrated to generate cold heat using a chemical heat pump.

Specifications

Molecular Formula CNa2O3 -+ 10H2O
Linear Formula Na2CO3 -+ 10H2O
Quantity 250 g
MDL Number MFCD00149178
Synonym Sal soda; Soda
Molecular Weight (g/mol) 286.14
Percent Purity 99.999% trace metals basis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.