Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium carbonate, Powder, ≥99.5%, ACS reagent, Solstice

Catalog No. 60045365 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
Packaging:
Fiberboard Box
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-365 500 g Poly Bottle
60-045-364 2.5 kg Poly Bottle
60-045-362 12 kg Fiberboard Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-045-365 Supplier Solstice Supplier No. 223530500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Powder,≥99.5%, ACS reagent

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 497-19-8
Molecular Formula CNa2O3
Molecular Weight (g/mol) 105.99
MDL Number MFCD00003494
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
Synonym sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
PubChem CID 10340
ChEBI CHEBI:29377
SMILES [Na+].[Na+].[O-]C([O-])=O

Specifications

Melting Point 851°C (literature)
CAS Min % ≥90.0000%
CAS Max % ≤100.0000%
Physical Form Powder
Quantity 500 g
Linear Formula Na2CO3
Beilstein 4154566
IUPAC Name disodium;carbonate
Formula Weight 105.99
Percent Purity >99.5%
Grade ACS Reagent
Packaging Poly Bottle
Density 2.53 g/cm3
Assay ex ignited substance
Chemical Name or Material Sodium carbonate, Powder, ≥99.5%, ACS reagent
Show More Show Less
ShelfLife 1260 days from date of manufacture
EINECSNumber 207-838-8
Recommended Storage Room Temp
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.