Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium carbonate, Puriss., meets analytical spec. of BP, Ph. Eur., NF, FCC, E500, anhydrous, 99.5-100.5% (calc. to the dried substance), Solstice

Catalog No. 6001983 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-019-83 5 kg
60-019-81 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-019-83 Supplier Solstice Supplier No. 134185KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Puriss., meets analytical specification of BP, Ph. Eur., NF, FCC, E500, anhydrous, 99.5-100.5% (calc. to the dried substance)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 497-19-8
Molecular Formula CNa2O3
Molecular Weight (g/mol) 105.99
MDL Number MFCD00003494
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
Synonym sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
PubChem CID 10340
ChEBI CHEBI:29377
SMILES [Na+].[Na+].[O-]C([O-])=O

Specifications

Physical Form Anhydrous
Quantity 5 kg
Linear Formula Na2CO3
UN Number NONH for all modes of transport
Beilstein 4154566
IUPAC Name disodium;carbonate
Formula Weight 105.99g/mol
Percent Purity 99.5 to 100.5% (calc. to the dried substance)
Grade puriss.
Chemical Name or Material Sodium Carbonate, Meets analytical specification of Ph.Eur., BP, NF, FCC
Hazard Statement P305 + P351 + P338
Precautionary Statement H319
EINECSNumber 207-838-8
RTECSNumber VZ4050000
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.