Learn More
Hach Company Sodium Carbonate Solution, 100 mL MDB
Supplier:  Hach Company 67532
Encompass
For determination of Tannin and Lignin by the Tyrosine method. Hach method 8193 100 mL Marked Dropping Bottle
Catalog No. 50-199-4423