Learn More
Hach Company Sodium Carbonate Solution, 500 mL
Supplier:  Hach Company 67549
Encompass
For determination of Tannin and Lignin by the Tyrosine method. Hach method 8193 500 mL bottle
Catalog No. 50-199-4424