Learn More
Medchemexpress LLC Sodium carboxyl methylstarch | 9063-38-1 | C2H4O3·xNa·x | 50 G
SDP
Catalog No. 5000283426
Encompass_Preferred
Sodium carboxyl methylstarch is the sodium salt of carboxyl methylstarch, a modified starch used as a biochemical reagent and pharmaceutical excipient. It is supplied as a white to off-white solid intended for laboratory research use only and commonly formulated as a disintegrant or viscosity modifier in life science applications.
  • White to off-white solid appearance.
  • Sodium salt of carboxyl methylstarch.
  • Used as a biochemical reagent and pharmaceutical excipient.
  • Common applications include tablet disintegration and formulation testing.
  • Available in laboratory pack sizes (25 g, 50 g, 100 g).
  • CAS number 9063-38-1; for research use only.
Catalog No. 50-002-83426 Supplier Medchemexpress LLC Supplier No. HYW13396350G
Sodium carboxyl methylstarch is the sodium salt of carboxyl methylstarch, a modified starch used as a biochemical reagent and pharmaceutical excipient. It is supplied as a white to off-white solid intended for laboratory research use only and commonly formulated as a disintegrant or viscosity modifier in life science applications.
  • White to off-white solid appearance.
  • Sodium salt of carboxyl methylstarch.
  • Used as a biochemical reagent and pharmaceutical excipient.
  • Common applications include tablet disintegration and formulation testing.
  • Available in laboratory pack sizes (25 g, 50 g, 100 g).
  • CAS number 9063-38-1; for research use only.