Learn More
Medchemexpress LLC Sodium carboxymethyl cellulose | 9004-32-4 | 100mg
SDP
Catalog No. 5000268646
Encompass_Preferred
Sodium carboxymethyl cellulose (Viscosity 800-1200 mPa s) is the sodium salt of cellulose arboxymethyl and frequently used as viscous agent paste and barrier agent
Catalog No. 50-002-68646 Supplier Medchemexpress LLC Supplier No. HYY0703100MG
Sodium carboxymethyl cellulose (Viscosity 800-1200 mPa s) is the sodium salt of cellulose arboxymethyl and frequently used as viscous agent paste and barrier agent