Learn More
Medchemexpress LLC Sodium carboxymethyl cellulose | 9004-32-4 | MFCD00081472 | 1g
SDP
Catalog No. 5000266362
Encompass_Preferred
Sodium carboxymethyl cellulose (CMC-Na) (MW 250000) is a sodium salt of carboxymethyl cellulose Sodium carboxymethyl cellulose has adsorption and corrosion inhibition on low-carbon steel in an acidic medium Sodium carboxymethyl cellulose can be used as a thickener paste and barrier agent[1][2][3]
Catalog No. 50-002-66362 Supplier Medchemexpress LLC Supplier No. HYY1889A1G
Sodium carboxymethyl cellulose (CMC-Na) (MW 250000) is a sodium salt of carboxymethyl cellulose Sodium carboxymethyl cellulose has adsorption and corrosion inhibition on low-carbon steel in an acidic medium Sodium carboxymethyl cellulose can be used as a thickener paste and barrier agent[1][2][3]