Learn More
Medchemexpress LLC Sodium carboxymethyl cellulose | 9004-32-4 | MFCD00081472 | 1g
SDP
Catalog No. 5000266192
Encompass_Preferred
Sodium carboxymethyl cellulose (Viscosity 5000-15000 mPa s) is the sodium salt of cellulose arboxymethyl and frequently used as viscous agent paste and barrier agent
Catalog No. 50-002-66192 Supplier Medchemexpress LLC Supplier No. HYY18891G
Sodium carboxymethyl cellulose (Viscosity 5000-15000 mPa s) is the sodium salt of cellulose arboxymethyl and frequently used as viscous agent paste and barrier agent