Learn More
Medchemexpress LLC Sodium carboxymethyl cellulose | 9004-32-4 | MFCD00081472 | 99.0% | 263.20 g/mol | C8H16NaO8 | 50g
SDP
Catalog No. 5000268799
Encompass_Preferred
Sodium carboxymethyl cellulose (Viscosity 300-800mpa s) is the sodium salt of cellulose arboxymethyl and frequently used as viscous agent paste and barrier agent
Catalog No. 50-002-68799 Supplier Medchemexpress LLC Supplier No. HYY0703C50G
Sodium carboxymethyl cellulose (Viscosity 300-800mpa s) is the sodium salt of cellulose arboxymethyl and frequently used as viscous agent paste and barrier agent