Learn More
Medchemexpress LLC Sodium carboxymethyl cellulose | 9004-32-4 | MFCD00081472 | 99.6% | 263.20 g/mol | C8H16NaO8 | 500mg
SDP
Catalog No. 5000267784
Encompass_Preferred
Sodium carboxymethyl cellulose (Viscosity 800-1200 mPa s) is the sodium salt of cellulose arboxymethyl and frequently used as viscous agent paste and barrier agent
Catalog No. 50-002-67784 Supplier Medchemexpress LLC Supplier No. HYY0703500MG
Sodium carboxymethyl cellulose (Viscosity 800-1200 mPa s) is the sodium salt of cellulose arboxymethyl and frequently used as viscous agent paste and barrier agent