Learn More
Medchemexpress LLC Sodium carboxymethyl cellulose | 9004-32-4 | MFCD00081472 | 99.8% | [C6H7O2(OH)x(OCH2COONa)y]n | 500g
SDP
Catalog No. 5000269793
Encompass_Preferred
Sodium carboxymethyl cellulose (Viscosity 1200-1400 mPa s) is the sodium salt of cellulose arboxymethyl and frequently used as viscous agent paste and barrier agent
Catalog No. 50-002-69793 Supplier Medchemexpress LLC Supplier No. HYY0703A500G
Sodium carboxymethyl cellulose (Viscosity 1200-1400 mPa s) is the sodium salt of cellulose arboxymethyl and frequently used as viscous agent paste and barrier agent