Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Sodium Chlorate
SCI_ED

Catalog No. S25540A Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
2 kg
Packaging:
Lab Grade - Crystals
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25540A 2 kg Lab Grade - Crystals
S25540 500 g Lab Grade - Crystals
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25540A
Add to Cart
Add to Cart

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 7775-09-9
Molecular Formula ClNaO3
Molecular Weight (g/mol) 106.44
InChI Key YZHUMGUJCQRKBT-UHFFFAOYSA-M
Synonym sodium chlorate, chloric acid, sodium salt, asex, chlorate de sodium, chlorsaure, tumbleleaf, atlacide, desolet, kusatol, rasikal
PubChem CID 516902
ChEBI CHEBI:65242
IUPAC Name sodium chlorate
SMILES [Na+].[O-][Cl](=O)=O

Specifications

Color White
Quantity 2 kg
Assay Percent Range >99%
Linear Formula NaClO3
Formula Weight 106.44
Grade Laboratory
Packaging Lab Grade - Crystals
Density 2.49g/mL
Chemical Name or Material Sodium Chlorate
Safety handling Check Alert storage code Red
ShelfLife 5 years
DOTInformation DOT Class 5.1, II, : Oxidizer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.