Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Chloride, 0.0250 N (N/40), Ricca Chemical
SDP

Catalog No. R72220004A Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R72220004A 4 L Natural Poly Bottle
7222-16 500 mL Natural Poly Bottle
7222-32 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. R72220004A Supplier Ricca Chemical Company Supplier No. 72221
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7647-14-5 , 7732-18-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

Color Colorless
Assay Percent Range 0.02500 ± 0.00001 N at 20°C
Solubility Information Solubility in Water: Infinite
Grade Laboratory
Odor Odorless
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
pH 6.7 to 7.3
Quantity 4 L
Physical Form Liquid
Chemical Name or Material Sodium Chloride, 0.0250 Normal (N/40)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.