Learn More
McKesson General Medical SODIUM CHLORIDE 0. DILUENT Sodium Chloride 0.9% Injection
Catalog No. NC1763998
Encompass_Preferred
Single dose plastic flip-top vial
This preparation is designed solely for parenteral use only after addition of drugs that require dilution or must be dissolved in an aqueous vehicle prior to injection
It contains no bacteriostat, antimicrobial agent or added buffer and is supplied only in single-dose containers to dilute or dissolve drugs for injection
A medical license and/or supplier forms may need to be completed in order to purchase this item
Catalog No. NC1763998 Supplier McKesson General Medical Supplier No. 239940
Single dose plastic flip-top vial
This preparation is designed solely for parenteral use only after addition of drugs that require dilution or must be dissolved in an aqueous vehicle prior to injection
It contains no bacteriostat, antimicrobial agent or added buffer and is supplied only in single-dose containers to dilute or dissolve drugs for injection
A medical license and/or supplier forms may need to be completed in order to purchase this item