Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Chloride, 1% w/v Standardized, Reagents

Catalog No. 01333655
Change view
Click to view available options
Quantity: 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-333-655 Quantity: 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-333-655 Supplier Reagents Holdings Llc Supplier No. CS1417004A
Add to Cart
Add to Cart

Sodium Chloride, 1% w/v Standardized, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Sodium Chloride, 1% w/v Standardized
Boiling Point 100°C
Color Colorless
CAS 7732-18-5, 7647-14-5
pH 6.7 to 7.3
Physical Form Liquid
Quantity 4 L
Molecular Formula NaCl/H2O
Synonym Table salt solution
Percent Purity 1%
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.0071
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.