Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Chloride, 10% (w/v), Ricca Chemical
SDP

Catalog No. 01213117 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-213-117 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-213-117 Supplier Ricca Chemical Company Supplier No. R7217000500A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7732-18-5 , 7647-14-5 , 7647-14-5 , 7732-18-5
Molecular Weight (g/mol) 58.44 g/mol

Specifications

Boiling Point Approximately 100°C
Melting Point Approximately 0°C
Color Colorless
CAS Min % 90.64%, 9.36%
Physical Form Liquid
Quantity 500 mL
CAS Max % 9.36%, 90.64%
Percent Purity 10% W/V
Grade Laboratory
Packaging 500 mL HDPE Bottle
Appearance Colorless Liquid
Chemical Name or Material Sodium Chloride
Show More Show Less
ShelfLife 24 Months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.