Learn More
Hach Company Conductivity Standard Solution, 1000 µS/cm,NaCl, 50 mL
Supplier:  Hach Company 1440026
Encompass
Conductivity standard solution,1000 µS/cm ±10 µS/cm at 25°C, 491 mg/L ±2.5 mg/L as Sodium Chloride (NaCl), (1 µS/cm ~ 0.5 mg/L TDS), 50 mL pour-top bottle.
Catalog No. 50-199-5021