Learn More
Hach Company Sodium Chloride Standard Solution, 10,000 µS/cm, 500 mL
Supplier:  Hach Company 2972249
Encompass
For accuracy checks in conductivity measurements. 5.37 g/L as NaCl. 10,000 µS/cm. 500 mL bottle.
Catalog No. 50-199-6978