Learn More
Hach Company Conductivity Standard Solution, 10000 µS/cm, NaCl, 50 mL
Catalog No. 501996977 Shop All Hach Company Products
Encompass
Conductivity standard solution,1000 µS/cm ±100 µS/cm at 25°C, 5.37 g/L ±0.03 g/L as Sodium Chloride (NaCl), (1 µS/cm ~ 0.5 mg/L TDS), 50 mL pour-top bottle.
Catalog No. 50-199-6977 Supplier Hach Company Supplier No. 2972226
Conductivity standard solution,1000 µS/cm ±100 µS/cm at 25°C, 5.37 g/L ±0.03 g/L as Sodium Chloride (NaCl), (1 µS/cm ~ 0.5 mg/L TDS), 50 mL pour-top bottle.