Learn More
Hach Company Conductivity Standard Solution, 1990 µS/cm, NaCl, 100 mL
Catalog No. 501994680 Shop All Hach Company Products
Encompass
Conductivity standard solution,1990 µS/cm ±20 µS/cm at 25°C, 1000 mg/L as Sodium Chloride (NaCl), (1 µS/cm ~ 0.5 mg/L TDS), 100 mL bottle.
Catalog No. 50-199-4680 Supplier Hach Company Supplier No. 210542
Conductivity standard solution,1990 µS/cm ±20 µS/cm at 25°C, 1000 mg/L as Sodium Chloride (NaCl), (1 µS/cm ~ 0.5 mg/L TDS), 100 mL bottle.