Learn More
Hach Company Sodium Chloride Standard Solution, 491 mg/L NaCl (1000 µS/cm), 500 mL
Catalog No. 501995023 Shop All Hach Company Products
Encompass
For standardization and accuracy checks in conductivity measurements
Catalog No. 50-199-5023 Supplier Hach Company Supplier No. 1440049
For standardization and accuracy checks in conductivity measurements