Learn More
Hach Company Conductivity Standard Solution, 100 µS/cm, NaCl, 50 mL
Supplier:  Hach Company 2971826
Encompass
Conductivity standard solution,100 µS/cm ±1 µS/cm at 25°C, 49 mg/L ±0.25 mg/L as Sodium Chloride (NaCl), (1 µS/cm ~ 0.5 mg/L TDS), 50 mL pour-top bottle.
Catalog No. 50-199-6974