Learn More
Hach Company Conductivity Standard Solution, 18000 µS/cm, NaCl, 100 mL
Catalog No. 501995764 Shop All Hach Company Products
Encompass
Conductivity standard solution,18000 µS/cm ±50 µS/cm at 25°C, 10246 mg/L ±100 mg/L as Sodium Chloride (NaCl), (1 µS/cm ~ 0.5 mg/L TDS), 100 mL bottle.
Catalog No. 50-199-5764 Supplier Hach Company Supplier No. 2307442
Conductivity standard solution,18000 µS/cm ±50 µS/cm at 25°C, 10246 mg/L ±100 mg/L as Sodium Chloride (NaCl), (1 µS/cm ~ 0.5 mg/L TDS), 100 mL bottle.