Learn More
Hach Company Sodium Chloride Standard Solution, 18,000 µS/cm, 500 mL
Catalog No. 501995765 Shop All Hach Company Products
Encompass
Conductivity Standard. 18,000 µS/cm (10,246 mg/L NaCl). 500 mL.
Catalog No. 50-199-5765 Supplier Hach Company Supplier No. 2307449
Conductivity Standard. 18,000 µS/cm (10,246 mg/L NaCl). 500 mL.