Learn More
Boston Bioproducts Inc Sodium Chloride (1 M) - 500ml
Catalog No. NC2372941
Encompass
Sodium Chloride, also refered to as saline, is a solution consisting of dissolved sodium chloride salt in varying sodium (Na+) and Chloride (Cl-) concentrations. This solution creates isotonic environments that mimic saline concentrations within living organisms. Sodium chloride is commonly used to increase the ionic strength in buffer solutions for molecular biology applications, cell culture and biochemical assays. This product is formulated at a 1M concentration.
Catalog No. NC2372941 Supplier Boston Bioproducts Inc Supplier No. BM247500ML
Sodium Chloride, also refered to as saline, is a solution consisting of dissolved sodium chloride salt in varying sodium (Na+) and Chloride (Cl-) concentrations. This solution creates isotonic environments that mimic saline concentrations within living organisms. Sodium chloride is commonly used to increase the ionic strength in buffer solutions for molecular biology applications, cell culture and biochemical assays. This product is formulated at a 1M concentration.