Learn More
Hach Company Sodium Chloride Standard Solution, 491 mg/L NaCl (1000 µS/cm), 100 mL
Catalog No. 501995022 Shop All Hach Company Products
Encompass
For calibrations/accuracy checks on conductivity meters.
Catalog No. 50-199-5022 Supplier Hach Company Supplier No. 1440042
For calibrations/accuracy checks on conductivity meters.