Learn More
Hach Company Sodium Chloride Standard Solution, 491mg/L NaCl (1000 µS/cm), 1L
Catalog No. 501995024 Shop All Hach Company Products
Encompass
For standardization and accuracy checks in conductivity measurements
Catalog No. 50-199-5024 Supplier Hach Company Supplier No. 1440053
For standardization and accuracy checks in conductivity measurements