Learn More
Hach Company Conductivity Standard Solution, 180 µS/cm, NaCl, 100 mL
Supplier:  Hach Company 2307542
Encompass
Conductivity standard solution,180 µS/cm ±10 µS/cm at 25°C, 85.47 mg/L as Sodium Chloride (NaCl), (1 µS/cm ~ 0.5 mg/L TDS), 100 mL bottle.
Catalog No. 50-199-5766