Learn More
Hach Company Sodium Chloride Standard Solution, 85.47 mg/L (180 µS/cm), 1L
Catalog No. 501995767 Shop All Hach Company Products
Encompass
For standardization and accuracy checks in conductivity measurements
Catalog No. 50-199-5767 Supplier Hach Company Supplier No. 2307553
For standardization and accuracy checks in conductivity measurements