Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium chloride, 99.5%, for analysis

Catalog No. p-3735103
Change view
Click to view available options
Quantity:
1 kg
5 kg
25 kg
Packaging:
Plastic Bottle
Plastic Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC207790010 1 kg Plastic Bottle
AC207790050 5 kg Plastic Bottle
AC207790250 25 kg Plastic Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC207790010 Supplier Thermo Scientific Chemicals Supplier No. 207790010
Only null left
Add to Cart

Sodium chloride, 99.5%, ClNa, CAS Number-7647-14-5, sodium chloride nacl, salt, common salt, rock salt, halite, purex, table salt, sodium chloride, saline, dendritis, 1kg, 99.4% min. (Argentometry), 10ppm max., 1461.0 deg.C, 50ppm max., 20ppm max., CHEBI:26710, White, 231-598-3, 13,280, 58.44 | CAS: 7647-14-5 | ClNa | 58.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium;chloride
SMILES [Na+].[Cl-]

Specifications

Boiling Point 1461.0°C
Melting Point 801.0°C
Color White
pH 5.0 to 8.0 (5% soln. at 20°C)
Physical Form Crystalline Powder
Quantity 1 kg
Assay Percent Range 99.4% min. (Argentometry)
Linear Formula NaCl
Fieser 13,280
Merck Index 15, 8734
Solubility Information Solubility in water: 360g/L (20°C). Other solubilities: soluble in glycerol (1g/10mL), ammonia, slightly soluble in alcohol, nearly insoluble in hydrochloric acid
Formula Weight 58.44
Percent Purity 99.5%
Grade Analytical
Packaging Plastic Bottle
Chemical Name or Material Sodium chloride
Show More Show Less
EINECSNumber 231-598-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.