Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium chloride, 99.99%, Suprapur™, MilliporeSigma™

Catalog No. M1064060500 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1064060500 500 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1064060500 Supplier MilliporeSigma Supplier No. 1.06406.0500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

Color Colorless
Physical Form Solid
Quantity 500 g
Assay Percent Range 99.99%
Linear Formula NaCl
Solubility Information 358g/L (20°C)
Formula Weight 58.44
Grade Suprapur
Packaging Plastic Bottle
Vapor Pressure 1.3 hPa (865°C)
Density 2.17g/cm3 (20°C)
Chemical Name or Material Sodium chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.