Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Chloride, ACS, ≥99.5%, Solstice

Catalog No. 60047483 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
6 x 500 g
4 x 2.5 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-047-483 500 g
60-047-480 2.5 kg
60-047-486 6 x 500 g
60-047-482 4 x 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-047-483 Supplier Solstice Supplier No. 746398500G
Add to Cart
Add to Cart

Free-flowing, ACS reagent, anhydrous,≥99%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
SMILES [Na+].[Cl-]

Specifications

Boiling Point 1465°C
Melting Point 800°C
Color Colorless
pH 7.0 (at 20°C)
Physical Form Crystalline
Quantity 500 g
Linear Formula NaCl
UN Number NONH for all modes of transport
Beilstein 3534976
Solubility Information 358g/L at 20°C
IUPAC Name sodium;chloride
Formula Weight 58.44g/mol
Percent Purity ≥99%
Odor Odorless
Packaging Poly bottle
Density 2.160g/cm3 at 20°C
Chemical Name or Material Sodium chloride
Show More Show Less
RTECSNumber 231-598-3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.