Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Chloride, ACS Reagent Grade, Ricca Chemical
SDP

Catalog No. RDCS043012F Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDCS043012F 12 kg
RDCS0430500 500 g
RDCS043025A 2.5 kg
RDCS043050F 50 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. RDCS043012F Supplier Ricca Chemical Company Supplier No. RDCS043012F4
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
IUPAC Name sodium chloride

Specifications

pH min 99%
CAS Max % 100
Physical Form Adhering Powder and/or Lumps
Quantity 12 kg
UN Number min 99%
Synonym min 99%
Solubility Information min 99%
SMILES min 99%
PubChem CID min 99%
Percent Purity min 99%
Grade min 99%
Packaging Fiber Drum
Product Line min 99%
Refractive Index min 99%
Residue after Ignition min 99%
Sensitivity min 99%
Specific Gravity min 99%
Vapor Pressure min 99%
Chemical Name or Material Sodium Chloride
Show More Show Less
ShelfLife min 99%
RTECSNumber min 99%
TSCA min 99%
storageNote1 min 99%
Recommended Storage min 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.