Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Chloride, ACS Reagent, Reagents

Catalog No. 01337744
Change view
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
12 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-744 10 kg
01-337-743 12 kg
01-337-742 2.5 kg
01-337-740 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01-337-744 Supplier Reagents Holdings Llc Supplier No. C131300010KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium Chloride, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym Table salt
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

Boiling Point 1461°C
Melting Point 801°C
Color Colorless/White
pH 6.7 to 7.3
Physical Form Solid
Quantity 10 kg
Grade ACS
Packaging Poly Pail
Specific Gravity 2.17
Chemical Name or Material Sodium Chloride, ACS Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.