Learn More
NEOGEN Sodium Chloride (Bacteriological), 500 g
SDP
Catalog No. 502015840
Encompass_Preferred
Sodium Chloride is a salt often used when preparing microbiological culture media in a laboratory setting. Sodium Chloride is not intended for use in the diagnosis of disease or other conditions in humans. When added to culture media Sodium Chloride allows for adjustments in osmotic pressure.For more information visit https://www.neogen.com/solutions/Microbiology/sodium-chloride-bacteriological
Catalog No. 50-201-5840 Supplier NEOGEN Supplier No. 700004698
Sodium Chloride is a salt often used when preparing microbiological culture media in a laboratory setting. Sodium Chloride is not intended for use in the diagnosis of disease or other conditions in humans. When added to culture media Sodium Chloride allows for adjustments in osmotic pressure.For more information visit https://www.neogen.com/solutions/Microbiology/sodium-chloride-bacteriological